siRNA Details
| Gene Symbol: E130307A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087502.1 | - | 2 | 290 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130307A14Rik| Gene Symbol: E130307A14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087502.1 | - | 2 | 290 | ||