siRNA Details
| Gene Symbol: Cyb5d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045525.1 | NP_001038990.1 | 3 | 397 | ||
| GenBank mRNA | BC099556.1 | - | 3 | 397 | 30915659 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyb5d1| Gene Symbol: Cyb5d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045525.1 | NP_001038990.1 | 3 | 397 | ||
| GenBank mRNA | BC099556.1 | - | 3 | 397 | 30915659 | |