siRNA Details
| Gene Symbol: Prps1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037746.3 | NP_001032835.2 | 2 | 3658 | ||
| GenBank mRNA | AK089022.1 | - | 1 | 687 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prps1l3| Gene Symbol: Prps1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037746.3 | NP_001032835.2 | 2 | 3658 | ||
| GenBank mRNA | AK089022.1 | - | 1 | 687 | ||