siRNA Details
| Gene Symbol: 9330182O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256056.1 | NP_001242985.1 | 4 | 621 | ||
| GenBank mRNA | AK034361.1 | - | 4 | 623 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330182O14Rik| Gene Symbol: 9330182O14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256056.1 | NP_001242985.1 | 4 | 621 | ||
| GenBank mRNA | AK034361.1 | - | 4 | 623 | ||