siRNA Details
| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177375.1 | NP_796349.1 | 9 | 1311 | ||
| GenBank mRNA | AK043912.1 | - | 8 | 1546 | ||
| BC094931.1 | - | 9 | 1221 | 6436242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab26| Gene Symbol: Rab26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177375.1 | NP_796349.1 | 9 | 1311 | ||
| GenBank mRNA | AK043912.1 | - | 8 | 1546 | ||
| BC094931.1 | - | 9 | 1221 | 6436242 | ||