siRNA Details
| Gene Symbol: Treml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033405.2 | NP_001028577.2 | 5 | 3899 | ||
| GenBank mRNA | AK037593.1 | - | 5 | 3374 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Treml2| Gene Symbol: Treml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033405.2 | NP_001028577.2 | 5 | 3899 | ||
| GenBank mRNA | AK037593.1 | - | 5 | 3374 | ||