siRNA Details
| Gene Symbol: Mcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085373.1 | NP_001078842.1 | 19 | 7472 | ||
| NM_001085374.1 | NP_001078843.1 | 17 | 7440 | |||
| GenBank mRNA | AK162662.1 | - | 1 | 4490 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mcc| Gene Symbol: Mcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085373.1 | NP_001078842.1 | 19 | 7472 | ||
| NM_001085374.1 | NP_001078843.1 | 17 | 7440 | |||
| GenBank mRNA | AK162662.1 | - | 1 | 4490 | ||