siRNA Details
| Gene Symbol: Gm826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033411.3 | NP_001028583.1 | 3 | 678 | ||
| GenBank mRNA | AK142710.1 | - | 3 | 679 | ||
| BC147270.1 | - | 3 | 626 | 9056346 | ||
| BC147271.1 | - | 3 | 626 | 9056326 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm826| Gene Symbol: Gm826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033411.3 | NP_001028583.1 | 3 | 678 | ||
| GenBank mRNA | AK142710.1 | - | 3 | 679 | ||
| BC147270.1 | - | 3 | 626 | 9056346 | ||
| BC147271.1 | - | 3 | 626 | 9056326 | ||