siRNA Details
| Gene Symbol: Gm14325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024849.2 | NP_001020020.2 | 4 | 2537 | ||
| GenBank mRNA | BC036144.1 | - | 1 | 1757 | ||
| BC092391.1 | - | 4 | 3121 | 6853907 | ||
| BC092392.1 | - | 5 | 2014 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14325| Gene Symbol: Gm14325 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024849.2 | NP_001020020.2 | 4 | 2537 | ||
| GenBank mRNA | BC036144.1 | - | 1 | 1757 | ||
| BC092391.1 | - | 4 | 3121 | 6853907 | ||
| BC092392.1 | - | 5 | 2014 | |||