siRNA Details
| Gene Symbol: Fnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162999.2 | NP_001156471.1 | 17 | 6051 | ||
| GenBank mRNA | AK052684.1 | - | 3 | 3501 | ||
| AK122512.1 | - | 9 | 5129 | |||
| AK141700.1 | - | 6 | 4175 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnip2| Gene Symbol: Fnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162999.2 | NP_001156471.1 | 17 | 6051 | ||
| GenBank mRNA | AK052684.1 | - | 3 | 3501 | ||
| AK122512.1 | - | 9 | 5129 | |||
| AK141700.1 | - | 6 | 4175 | |||