siRNA Details
| Gene Symbol: AI464131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085515.2 | NP_001078984.1 | 2 | 1791 | ||
| GenBank mRNA | AK173117.1 | - | 2 | 1773 | ||
| BC036141.1 | - | 1 | 541 | |||
| BC137640.1 | - | 2 | 1748 | 8860659 | ||
| BC140956.1 | - | 2 | 1748 | 9055568 | ||

AI464131
Genomic Map