siRNA Details
| Gene Symbol: Tmem120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039723.2 | NP_001034812.1 | 5 | 564 | ||
| GenBank mRNA | AK132283.1 | - | 5 | 533 | ||
| AK158123.1 | - | 5 | 523 | |||
| AK172110.1 | - | 5 | 504 | |||
| BC022593.1 | - | 5 | 515 | 4038363 | ||
| BC115785.1 | - | 4 | 408 | 40104955 | ||
| BC118014.1 | - | 5 | 525 | 40104954 | ||

Tmem120b
Genomic Map