siRNA Details
| Gene Symbol: Gal3st4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033416.2 | NP_001028588.1 | 3 | 2809 | ||
| GenBank mRNA | AK132558.1 | - | 3 | 2810 | ||
| AK138993.1 | - | 4 | 1474 | |||
| AK162468.1 | - | 1 | 1034 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gal3st4| Gene Symbol: Gal3st4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033416.2 | NP_001028588.1 | 3 | 2809 | ||
| GenBank mRNA | AK132558.1 | - | 3 | 2810 | ||
| AK138993.1 | - | 4 | 1474 | |||
| AK162468.1 | - | 1 | 1034 | |||