siRNA Details
| Gene Symbol: Thsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164805.1 | NP_001158277.1 | 20, 21 | 4189 | ||
| GenBank mRNA | AK133324.1 | - | 15, 16 | 2269 | ||
| AK173072.1 | - | 20, 21 | 3960 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Thsd7a| Gene Symbol: Thsd7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164805.1 | NP_001158277.1 | 20, 21 | 4189 | ||
| GenBank mRNA | AK133324.1 | - | 15, 16 | 2269 | ||
| AK173072.1 | - | 20, 21 | 3960 | |||