siRNA Details
| Gene Symbol: D630045J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194061.2 | NP_918950.2 | 20 | 9141 | ||
| GenBank mRNA | AK141785.1 | - | 1 | 514 | ||
| BC042666.1 | - | 1 | 2504 | |||
| BC059853.1 | - | 1 | 2490 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D630045J12Rik| Gene Symbol: D630045J12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194061.2 | NP_918950.2 | 20 | 9141 | ||
| GenBank mRNA | AK141785.1 | - | 1 | 514 | ||
| BC042666.1 | - | 1 | 2504 | |||
| BC059853.1 | - | 1 | 2490 | |||