siRNA Details
| Gene Symbol: Cdon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017358.1 | NP_059054.1 | 3, 4 | 530 | ||
| XM_006242791.2 | XP_006242853.1 | 3, 4 | 568 | |||
| XM_006242792.2 | XP_006242854.1 | 4, 5 | 1157 | |||
| XM_006242793.2 | XP_006242855.1 | 3, 4 | 568 | |||
| XM_008766066.1 | XP_008764288.1 | 4, 5 | 1967 | |||
| GenBank mRNA | AF004840.1 | - | 3, 4 | 530 | ||

Cdon
Genomic Map