siRNA Details
| Gene Symbol: Tmem52b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081186.1 | NP_001074655.1 | 5 | 2166 | ||
| GenBank mRNA | AK085389.1 | - | 5 | 2174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem52b| Gene Symbol: Tmem52b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081186.1 | NP_001074655.1 | 5 | 2166 | ||
| GenBank mRNA | AK085389.1 | - | 5 | 2174 | ||