siRNA Details
Gene Symbol: Tmem52b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081186.1 | NP_001074655.1 | 5 | 1471 | ||
GenBank mRNA | AK085389.1 | - | 5 | 1477 | ||
AK085571.1 | - | 5 | 1413 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmem52b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081186.1 | NP_001074655.1 | 5 | 1471 | ||
GenBank mRNA | AK085389.1 | - | 5 | 1477 | ||
AK085571.1 | - | 5 | 1413 |