siRNA Details
| Gene Symbol: Chrm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017362.4 | NP_059058.1 | 1672 | |||
| GenBank mRNA | M22926.1 | - | 1672 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrm5| Gene Symbol: Chrm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017362.4 | NP_059058.1 | 1672 | |||
| GenBank mRNA | M22926.1 | - | 1672 | |||
Set Membership: |
|