siRNA Details
| Gene Symbol: Htra4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081187.3 | NP_001074656.1 | 9 | 1439 | ||
| GenBank mRNA | AK046614.1 | - | 9 | 1358 | ||
| BC132380.1 | - | 9 | 1399 | 40130657 | ||
| BC145842.1 | - | 9 | 1399 | 40131145 | ||

Htra4
Genomic Map