siRNA Details
| Gene Symbol: Drc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042715.3 | NP_001036180.2 | 3 | 414 | ||
| GenBank mRNA | AK076786.1 | - | 3 | 409 | ||
| AY352586.2 | - | 3 | 411 | |||
| BC117982.1 | - | 2 | 388 | |||
| BC117983.1 | - | 2 | 388 | |||

Drc7
Genomic Map