siRNA Details
| Gene Symbol: Gm16130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243265.1 | NP_001230194.1 | 5 | 1552 | ||
| GenBank mRNA | AK054504.1 | - | 5 | 1553 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16130| Gene Symbol: Gm16130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243265.1 | NP_001230194.1 | 5 | 1552 | ||
| GenBank mRNA | AK054504.1 | - | 5 | 1553 | ||