siRNA Details
Gene Symbol: Gm16130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243265.1 | NP_001230194.1 | 5 | 2382 | ||
GenBank mRNA | AK054504.1 | - | 5 | 2383 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm16130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243265.1 | NP_001230194.1 | 5 | 2382 | ||
GenBank mRNA | AK054504.1 | - | 5 | 2383 |