siRNA Details
Gene Symbol: Gm5127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033541.2 | NP_001028713.1 | 2, 3 | 232 | ||
GenBank mRNA | AK044840.1 | - | 3, 4 | 317 | ||
AK142438.1 | - | 2, 3 | 233 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033541.2 | NP_001028713.1 | 2, 3 | 232 | ||
GenBank mRNA | AK044840.1 | - | 3, 4 | 317 | ||
AK142438.1 | - | 2, 3 | 233 |