siRNA Details
| Gene Symbol: Gm5127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033541.2 | NP_001028713.1 | 4 | 1342 | ||
| GenBank mRNA | AK142438.1 | - | 4 | 1343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5127| Gene Symbol: Gm5127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033541.2 | NP_001028713.1 | 4 | 1342 | ||
| GenBank mRNA | AK142438.1 | - | 4 | 1343 | ||