siRNA Details
| Gene Symbol: Gm853 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034872.2 | NP_001030044.1 | 9 | 1386 | ||
| GenBank mRNA | AK143920.1 | - | 9 | 1387 | ||
| BC147394.1 | - | 8 | 1251 | 9056469 | ||
| BC147395.1 | - | 8 | 1251 | 9056458 | ||

Gm853
Genomic Map