siRNA Details
| Gene Symbol: Trim75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033429.2 | NP_001028601.1 | 2 | 2220 | ||
| GenBank mRNA | AK136018.1 | - | 2 | 2222 | ||
| AK162165.1 | - | 1 | 1063 | |||
| AK163251.1 | - | 1 | 1577 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim75| Gene Symbol: Trim75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033429.2 | NP_001028601.1 | 2 | 2220 | ||
| GenBank mRNA | AK136018.1 | - | 2 | 2222 | ||
| AK162165.1 | - | 1 | 1063 | |||
| AK163251.1 | - | 1 | 1577 | |||