siRNA Details
| Gene Symbol: Trim75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033429.2 | NP_001028601.1 | 1 | 41 | ||
| GenBank mRNA | AK136018.1 | - | 1 | 43 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim75| Gene Symbol: Trim75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033429.2 | NP_001028601.1 | 1 | 41 | ||
| GenBank mRNA | AK136018.1 | - | 1 | 43 | ||