siRNA Details
| Gene Symbol: A4gnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077424.2 | NP_001070892.1 | 3 | 1142 | ||
| GenBank mRNA | AK136425.1 | - | 3 | 1143 | ||
| BC115614.1 | - | 2 | 1033 | 40102739 | ||
| BC115615.1 | - | 2 | 1033 | 40102743 | ||

A4gnt
Genomic Map