siRNA Details
| Gene Symbol: A4gnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077424.2 | NP_001070892.1 | 3 | 2389 | ||
| GenBank mRNA | AK136425.1 | - | 3 | 2390 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A4gnt| Gene Symbol: A4gnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077424.2 | NP_001070892.1 | 3 | 2389 | ||
| GenBank mRNA | AK136425.1 | - | 3 | 2390 | ||