siRNA Details
| Gene Symbol: B020031M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033769.2 | NP_001028941.1 | 2 | 485 | ||
| GenBank mRNA | AK139881.1 | - | 2 | 486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B020031M17Rik| Gene Symbol: B020031M17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033769.2 | NP_001028941.1 | 2 | 485 | ||
| GenBank mRNA | AK139881.1 | - | 2 | 486 | ||