siRNA Details
| Gene Symbol: Gm15104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101501.1 | NP_001094971.1 | 1 | 255 | ||
| GenBank mRNA | BC099570.1 | - | 2 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15104| Gene Symbol: Gm15104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101501.1 | NP_001094971.1 | 1 | 255 | ||
| GenBank mRNA | BC099570.1 | - | 2 | 332 | ||