siRNA Details
| Gene Symbol: Fkbp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045528.1 | NP_001038993.1 | 6 | 575 | ||
| GenBank mRNA | AK049459.1 | - | 6 | 520 | ||
| AK049509.1 | - | 6 | 576 | |||
| AK082565.1 | - | 6 | 524 | |||
| AK085087.1 | - | 6 | 563 | |||
| AK161705.1 | - | 6 | 505 | |||
| BC060651.1 | - | 6 | 489 | |||
| BC137706.1 | - | 6 | 509 | 8860725 | ||

Fkbp15
Genomic Map