siRNA Details
| Gene Symbol: Kdm7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033430.4 | NP_001028602.2 | 16 | 2218 | ||
| GenBank mRNA | AK035529.1 | - | 7 | 1232 | ||
| AK129429.1 | - | 5 | 560 | |||
| AK136169.1 | - | 4 | 356 | |||
| AK136209.1 | - | 16 | 2057 | |||
| AK136238.1 | - | 16 | 2205 | |||
| BC007161.1 | - | 16 | 1914 | |||
| BC145216.1 | - | 16 | 2104 | |||
| BC145848.1 | - | 16 | 2108 | 40131151 | ||

Kdm7a
Genomic Map