siRNA Details
| Gene Symbol: Zfp703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101502.1 | NP_001094972.1 | 2 | 2223 | ||
| NM_001110508.1 | NP_001103978.1 | 3 | 2357 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp703| Gene Symbol: Zfp703 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101502.1 | NP_001094972.1 | 2 | 2223 | ||
| NM_001110508.1 | NP_001103978.1 | 3 | 2357 | |||