siRNA Details
| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033431.1 | NP_001028603.1 | 10 | 3431 | ||
| GenBank mRNA | AK149454.1 | - | 3 | 472 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp12| Gene Symbol: Nlrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033431.1 | NP_001028603.1 | 10 | 3431 | ||
| GenBank mRNA | AK149454.1 | - | 3 | 472 | ||