siRNA Details
| Gene Symbol: Spata22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045531.1 | NP_001038996.1 | 3 | 199 | ||
| GenBank mRNA | BC147586.1 | - | 3 | 181 | 9056576 | |
| BC147607.1 | - | 3 | 181 | 9056597 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata22| Gene Symbol: Spata22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045531.1 | NP_001038996.1 | 3 | 199 | ||
| GenBank mRNA | BC147586.1 | - | 3 | 181 | 9056576 | |
| BC147607.1 | - | 3 | 181 | 9056597 | ||