siRNA Details
Gene Symbol: Gm884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033434.2 | NP_001028606.2 | 1 | 1052 | ||
GenBank mRNA | AK029868.1 | - | 1 | 1052 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033434.2 | NP_001028606.2 | 1 | 1052 | ||
GenBank mRNA | AK029868.1 | - | 1 | 1052 |