siRNA Details
| Gene Symbol: Gm904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033770.1 | NP_001028942.1 | 2, 3 | 290 | ||
| GenBank mRNA | AK131725.1 | - | 2, 3 | 290 | ||
| BC147620.1 | - | 2, 3 | 280 | 9056610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm904| Gene Symbol: Gm904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033770.1 | NP_001028942.1 | 2, 3 | 290 | ||
| GenBank mRNA | AK131725.1 | - | 2, 3 | 290 | ||
| BC147620.1 | - | 2, 3 | 280 | 9056610 | ||