siRNA Details
| Gene Symbol: Lrch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033439.3 | NP_001028611.2 | 20 | 4553 | ||
| NM_001252132.1 | NP_001239061.1 | 19 | 4472 | |||
| GenBank mRNA | AK129264.1 | - | 20 | 4543 | ||
| AK160965.1 | - | 1 | 2289 | |||
| AK162105.1 | - | 9 | 2980 | |||
| BC028469.1 | - | 1 | 940 | |||

Lrch1
Genomic Map