siRNA Details
| Gene Symbol: Ptprs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019140.2 | NP_062013.1 | 8 | 1632 | ||
| GenBank mRNA | BC105753.1 | - | 1875 | 5599711 | ||
| L12329.1 | - | 8 | 1632 | |||
| L19180.1 | - | 1472 | ||||
| L19181.1 | - | 7 | 1472 | |||
| L19933.1 | - | 7 | 1546 | |||

Ptprs
Genomic Map