siRNA Details
| Gene Symbol: Gm1587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033440.2 | NP_001028612.1 | 3 | 778 | ||
| GenBank mRNA | AK139671.1 | - | 3 | 781 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1587| Gene Symbol: Gm1587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033440.2 | NP_001028612.1 | 3 | 778 | ||
| GenBank mRNA | AK139671.1 | - | 3 | 781 | ||