siRNA Details
| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201529.2 | NP_963287.2 | 14 | 2618 | ||
| GenBank mRNA | AK129231.1 | - | 11 | 2403 | ||
| AK137694.1 | - | 5 | 857 | |||
| BC049164.1 | - | 8 | 1203 | |||
| BC082553.1 | - | 12 | 2633 | |||
| BC145011.1 | - | 7 | 1021 | 9053559 | ||
| EF071857.1 | - | 14 | 2589 | |||

Lmo7
Genomic Map