siRNA Details
| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201529.2 | NP_963287.2 | 3, 4 | 439 | ||
| GenBank mRNA | BB614044.1 | - | 3, 4 | 455 | ||
| EF071857.1 | - | 3, 4 | 410 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmo7| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201529.2 | NP_963287.2 | 3, 4 | 439 | ||
| GenBank mRNA | BB614044.1 | - | 3, 4 | 455 | ||
| EF071857.1 | - | 3, 4 | 410 | |||