siRNA Details
| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201529.2 | NP_963287.2 | 31 | 5811 | ||
| GenBank mRNA | AK129231.1 | - | 27 | 5487 | ||
| BC082553.1 | - | 28 | 5717 | |||
| BC145011.1 | - | 24 | 4294 | 9053559 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmo7| Gene Symbol: Lmo7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201529.2 | NP_963287.2 | 31 | 5811 | ||
| GenBank mRNA | AK129231.1 | - | 27 | 5487 | ||
| BC082553.1 | - | 28 | 5717 | |||
| BC145011.1 | - | 24 | 4294 | 9053559 | ||