siRNA Details
| Gene Symbol: Gm933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256309.1 | NP_001243238.1 | 3 | 286 | ||
| GenBank mRNA | AI509129.1 | - | 3 | 286 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AA028844.1 | - | 286 | Chr. - - [-] on Build | ||

Gm933
Genomic Map