siRNA Details
| Gene Symbol: Zfp229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164676.1 | NP_001158148.1 | 6 | 1204 | ||
| GenBank mRNA | AK139846.1 | - | 6 | 995 | ||
| BC043476.1 | - | 3 | 795 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp229| Gene Symbol: Zfp229 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164676.1 | NP_001158148.1 | 6 | 1204 | ||
| GenBank mRNA | AK139846.1 | - | 6 | 995 | ||
| BC043476.1 | - | 3 | 795 | |||