siRNA Details
| Gene Symbol: Capn13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033444.2 | NP_001028616.1 | 1 | 33 | ||
| GenBank mRNA | AK136569.1 | - | 1 | 34 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Capn13| Gene Symbol: Capn13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033444.2 | NP_001028616.1 | 1 | 33 | ||
| GenBank mRNA | AK136569.1 | - | 1 | 34 | ||