siRNA Details
| Gene Symbol: Arl14epl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033446.2 | NP_001028618.1 | 5 | 2220 | ||
| GenBank mRNA | AK145795.1 | - | 5 | 2214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arl14epl| Gene Symbol: Arl14epl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033446.2 | NP_001028618.1 | 5 | 2220 | ||
| GenBank mRNA | AK145795.1 | - | 5 | 2214 | ||