siRNA Details
| Gene Symbol: Greb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083628.1 | NP_001077097.1 | 33 | 7497 | ||
| GenBank mRNA | AK220484.1 | - | 13 | 4014 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Greb1l| Gene Symbol: Greb1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083628.1 | NP_001077097.1 | 33 | 7497 | ||
| GenBank mRNA | AK220484.1 | - | 13 | 4014 | ||